Learn More
WTW™ Ammonia and Ammonium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 252027
1253.44 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11752245
  • 1290.00 DKK / Pack of 100
Explore more special offers
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For waste water, drinking water, sea water
Certificates
2 to 150 mg/L NH4-N, 2.6 to 193 mg/L NH4+
Cuvette
For PhotoLab™ S12, PhotoLab™ Spectral photometers
25 Pack
Photometric
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Ammonia and Ammonium Test Kits > Reagent Test; Detection Range: 2 to 150mg/L NH4-N, 2.6 to 193mg/L NH4+