Learn More
Ammonia buffer solution, Ammonium chloride / ammonia, pH 10, for complexometry, Honeywell Fluka™ Product Code.: 15644010
compliance-icons compliance-icons
Ammonia buffer solution, Ammonium chloride / ammonia, pH 10, for complexometry, Honeywell Fluka™
Product Code. p-7122060 Shop All Honeywell Fluka Products
1 L, 1 L
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
Product Code. 15644010
Brand: Honeywell-Fluka 335821L
Request Bulk or Custom Format
Ammonium chloride / ammonia, pH 10, for complexometry
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ammonia buffer solution
CAS 12125-02-9
Quantity 1 L
Molecular Formula ClH4N
MDL Number MFCD00011420
Packaging Poly bottle
UN Number NONH for all modes of transport
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name azanium;chloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonia buffer solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium chloride, Ammonia, aqueous solution
Product Content Correction
Product Title
Ammonia buffer solution, Ammonium chloride / ammonia, pH 10, for complexometry, Honeywell Fluka™ >