missing translation for 'onlineSavingsMsg'
Learn More

Ammonium perchlorate solution, Reagent Grade, ∼1 M in H2O, Honeywell™

Brand:  Honeywell Research Chemicals 09911-50L

Product Code. 16666263

  • 39041.19 DKK / 50 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

>100°C
<0°C
Colorless
≤ 90%
NH4ClO4
Reagent
Ammonium perchlorate solution
7732-18-5
>80%
50 L
117.49 g/mol
1.053 g/cm3 (20°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium perchlorate solution, Reagent Grade, ∼1 M in H2O, Honeywell™ > 50 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.