missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Peroxodisulfate, Extra Pure, Scharlab Product Code.: 11319843
compliance-icons compliance-icons compliance-icons

Ammonium Peroxodisulfate, Extra Pure, Scharlab

Product Code. 11319843
1 kg, 1kg
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11319843

Brand: Scharlab AM03701000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry and in the synthesis of polymers.

  • Synonyms: Ammonium persulfate, Peroxodisulfuric acid diammonium salt
  • Molecular Formula: (NH4)2S2O8
  • Molar Mass: 228.20g/mol
  • CAS: 7727-54-0
  • EIN ECS No.: 231-786-5

Specifications

Melting Point 120°C (decomposition)
CAS 7727-54-0
Color White to Yellow
Physical Form Powder
Quantity 1 kg
Molecular Formula H8N2O8S2
MDL Number MFCD00003390
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Weight (g/mol) 228.19
PubChem CID 62648
Formula Weight 228.20g/mol
Percent Purity ≥98% (iodometric)
Grade Extra Pure
Residue after Ignition 0.1% max (as SO4)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium peroxodisulfate, extra pure, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Peroxodisulfate, Extra Pure, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.