missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Synonyms: Sulfuric acid diammonium salt
- Molecular Formula: (NH4)2SO4
- Molar Mass: 132.14g/mol
- CAS: 7783-20-2
- EIN ECS No.: 231-984-1
Specifications
Specifications
Melting Point | 280°C (decomposition) |
CAS | 7783-20-2 |
pH | 4.5–6.0 (5%, water) |
Quantity | 1 kg |
Molecular Formula | H8N2O4S |
MDL Number | MFCD00003391 |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
SMILES | N.N.OS(O)(=O)=O |
IUPAC Name | sulfuric acid diamine |
Show More |
Safety and Handling

Product Identifier
- Ammonium sulfate, reagent grade, ACS, ISO, Reag. Ph Eur
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Sulfate, ACS Reagent Grade, Scharlab >
Spot an opportunity for improvement?Share a Content Correction