Learn More
Buerkle™ Spigot Stopcock™ Angled Product Code.: 10077970
Buerkle™ Spigot Stopcock™ Angled
Product Code. 10077970 Shop All Bürkle Products
6 to 8mm dia., Bottles with narrow openings, 1.905cm (0.75in) connection threads
Click to view available options
6 to 8mm dia., Bottles with narrow openings, 1.905cm (0.75in) connection threads
Not suitable for crystallising substances
Product Code. 10077970
Brand: Buerkle™ 05251000
Edge
Not suitable for crystallising substances
Discharge stopcock fits all containers with R 0.75 in. connection threads. Buerkle Angled Spigot Stopcock thread adapters facilitate connection to nearly all container types.
  • Tight-closing for complete seal
  • Effortless to use
  • With connecting cap nut, guaranteed vertical position of the cock
  • Pressure-resistant up to 2 bar at 20°C (without discharge bracket and grommet)
  • With discharge angle and rotatable spout preventing drips
  • With tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm
  • Media-contacting parts PP, PE and PTFE
Specifications
Includes R 0.75 in. Connection threads, connecting cap nut, discharge angle and rotatable tapered spoμt
Material Polypropylene, Polyethylene, PTFE
Product Type Stopcock Spigot
Sterility Non-sterile
  • Buerkle Angled Spigot StopCock
    Buerkle Angled Spigot StopCock
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Buerkle Angled Spigot Stopcock
    Product Content Correction
    Product Title
    Buerkle™ Spigot Stopcock™ Angled >