missing translation for 'onlineSavingsMsg'
Learn More

Agencinox™ Anodised Aluminum Colored Sterilization Boxes

Use these boxes for a variety of general laboratory applications. Agencinox™ Anodised Aluminum Colored Sterilization Boxes keep instruments safe and secure during sterilization process.

Brand:  Agencinox™ 8031

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11812692

  • 179.50 DKK / Each
Estimated Shipment: 08-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Anodized aluminum colored instrument boxes
  • Available in a variety of sizes
Specifications

Specifications

Natural
Rectangular
180 mm
40 mm
Aluminum
Sterilization Box
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers