Learn More
Agencinox™ Anodised Aluminum Colored Sterilization Boxes
Use these boxes for a variety of general laboratory applications. Agencinox™ Anodised Aluminum Colored Sterilization Boxes keep instruments safe and secure during sterilization process.
Brand: Agencinox™ 8030V
Additional Details : Weight : 0.80000kg
Description
- Anodized aluminum colored instrument boxes
- Available in a variety of sizes
Specifications
Green | |
Rectangular | |
180 mm | |
30 mm |
Aluminum | |
Sterilization Box | |
90 mm |