Learn More
Ansell TouchNTuff™ 73-500 Sterile Neoprene Cleanroom Gloves
Thin, sterile neoprene gloves offer added tactile sensitivity and provides chemical splash resistance against a broad range of chemicals. Compatible with Class 100/ ISO 5 cleanroom environments.
Brand: Ansell 73500080
5018.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Sensitive neoprene
- Ideal for double donning and dry applications
- Not made of natural rubber latex or accelerators
- Increased sensitivity due to the polymer thickness
- Anatomic hand specific design for comfort and fit
- Chemical splash resistance
- Long rolled beaded cuff
- Gamma irradiation sterilization
- Smooth grip, powder free
- Applications include:
- Blending of liquids
- Analytical testing/ measurements
- Blistering operations
- Pelletizing and pressing drugs
- Organic and other material synthesis
Specifications
AS/NZS 4179, ASTM D3577, ASTM D7160, ASTM D6978-05, ISO 10282, ISO 11193, ISO 13485 | |
Class 100 (ISO 5) | |
Cream | |
6.6 mil | |
Specific | |
Neoprene | |
0.15 mm | |
8 | |
6.6 mil | |
ISO 9001:2015 | |
TouchNTuff™ 73-500 | |
12 in. |
CE 0493, EN 420:2003 + A1:2009, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 421:2010 | |
0.65 | |
No Texture | |
0.16 mm | |
Yes | |
5.9 mil | |
Powder Free | |
Sterile | |
0.16 mm | |
73500080 | |
17 MPa | |
30.5 cm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2411028005 | Certificate of Irradiation | 04/06/2025 | 73500080 |
2502054805 | Certificate of Irradiation | 21/05/2025 | 73500080 |
2502001805 | Certificate of Irradiation | 19/05/2025 | 73500080 |
2411028005 | Certificate of Compliance | 04/06/2025 | 73500080 |
2502054805 | Certificate of Compliance | 21/05/2025 | 73500080 |