Learn More
anti-ATF4, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000468-AP41.1Set
Product Code. 16161764
  • 6445.00 DKK / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-ATF4, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-ATF4, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
ATF4
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ATF4.
CREB-2/CREB2/TAXREB67/TXREB
Mouse
RUO
468
Human
Liquid
ELISA
Biotin
ATF4
ATF4
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ATF4, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-ATF4, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only