missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD28 Matched Antibody Pair, Abnova™
Brand: Abnova H00000940-AP41.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-CD28 (100μg); Detection antibody: biotinylated mouse monoclonal anti-CD28, IgG2b Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
Host Species: Mouse, Mouse
Isotype: IgG2b, κ
Cross Reactivity: Human
ELISA Pair
Specifications
CD28 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD28. | |
MGC138290/Tp44 | |
Mouse | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD28 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD28, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Biotin | |
CD28 | |
CD28 | |
1 Set | |
940 | |
Human | |
IgG2b |