Learn More
CD3E, Human, Matched Antibody Pair (AP42), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000916-AP42.1Set
Product Code. 16191834
  • 6445.00 DKK / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-CD3E (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD3E
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD3E.
FLJ18683/T3E/TCRE
Mouse
RUO
916
Human
Liquid
ELISA
Biotin
CD3E
CD3E
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD3E (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only