Learn More
anti-HMBS Matched Antibody Pair, Abnova™
Brand: Abnova H00003145-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HMBS (100μg); Detection antibody: mouse purified polyclonal anti-HMBS, IgG2a κ (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMBS
Host Species: Rabbit, Mouse
Isotype: IgG, IgG2a κ
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
HMBS | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMBS. | |
PBG-D/PBGD/UPS | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HMBS (100 μg) 2. Detection antibody: mouse monoclonal anti-HMBS, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
HMBS | |
HMBS | |
Affinity Purified | |
RUO | |
3145 | |
Human | |
IgG, IgG2a κ |