Learn More
anti-IDO1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00003620-AP22.1Set
Product Code. 16192254
  • 6445.00 DKK / 1 set
Explore more special offers
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-IDO1 (100μg) , Detection antibody: mouse purified polyclonal anti-IDO1 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
IDO1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IDO1.
CD107B/IDO/INDO
Mouse, Rabbit
RUO
3620
Human
Liquid
ELISA
Unconjugated
IDO1
IDO1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IDO1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-IDO1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only