Learn More
anti-MYL5 Matched Antibody Pair, Abnova
Brand: Abnova H00004636-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Rabbit affinity purified polyclonal anti-MYL5 (100μg); Mouse monoclonal anti-MYL5, IgG2aκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.
ELISA
Specifications
MYL5 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human MYL5. | |
MYL5 | |
Affinity Purified | |
RUO | |
4636 | |
Human | |
IgG2a |
ELISA | |
Unconjugated | |
MYL5 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-MYL5 (100 μg) 2. Detection antibody: mouse monoclonal anti-MYL5, IgG2a Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |