missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTRK2, Human, Matched Antibody Pair (AP52), Abnova™
Brand: Abnova H00004915-AP52.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-NTRK2 (100μg); Detection antibody: rabbit purified polyclonal anti-NTRK2 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NTRK2.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
NTRK2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NTRK2. | |
GP145-TrkB/TRKB | |
Mouse, Rabbit | |
RUO | |
4915 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
NTRK2 | |
NTRK2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NTRK2 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-NTRK2 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |