Learn More
anti-TRIP10, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00009322-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16061379
  • 6285.00 DKK / 1 set
Estimated Shipment: 30-08-2024
to see stock.
Includes: Antibody pair for IP: mouse monoclonal anti-TRIP10 (300μg), Antibody pair for WB: rabbit polyclonal anti-TRIP10 (50μL)
Description
Description
Specifications
Specifications
TRIP10
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
CIP4/HSTP/STOT/STP
Mouse, Rabbit
RUO
9322
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
TRIP10
TRIP10
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-TRIP10 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-TRIP10 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only