missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 7 BshTI

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN0074

 View more versions of this product

Product Code. 15542430

  • 1290.00 DKK / 250 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
This item is not returnable. View return policy

Description

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 μL Anza 7 BshTI
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)

Store at -20°C.
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
250 units
Traditional Cloning
Approved for shipment on Wet or Dry Ice
BshTI
No
No
Restriction Enzyme Cloning System
Anza™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2836275 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberIVGN0074
Lot Number2859799 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberIVGN0074
Lot Number3028533 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberIVGN0074
Lot Number3095394 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberIVGN0074
Lot Number3094827 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog NumberIVGN0074
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Anza™ 7 BshTI > 25μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.