Learn More
Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)
Dissolved in glass-distilled water and titrated to pH 7.0 with NaOH
Brand: Applied Biosystems™ 4303442
This item is not returnable.
View return policy
Description
Each vial of GeneAmp deoxynucleotide triphosphates consists of 1mL of a 10mM blend of 2.5mM each of dATP, dCTP, dGTP, and dTTP.
- dNTPs are of high purity,making them for use in PCR reactions or other applications
- Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument system
PCR, PCR and Real-Time PCR
Specifications
GeneAmp™ dNTP Blend contains 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP (a blend of 10 mM dNTPs). Sufficient for 200 PCR amplifications, 50 μL each. Store at -20°C in a constant temperature freezer. | |
GeneAmp™ | |
12 x 1 mL | |
10 mM |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Application Notes & Tutorials: GeneAmp® 10 mM dNTP Mix with dTTP
File size: 17.1 kB
Application Notes & Tutorials: GeneAmp® 12.5 mM dNTP Mix with dUTP
File size: 20.8 kB
Product Content Correction
Product Title
Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea) > 12 x 1mL