Learn More
Applied Biosystems™ Nuclease-free Water (1.75 mL/tube) Product Code.: 10117584
Applied Biosystems™ Nuclease-free Water (1.75 mL/tube)
Product Code. 10117584
1 x 1.75 mL tube, Each
Click to view available options
Quantity:
1 x 1.75 mL tube
Unit Size:
Each
Product Code. 10117584
Brand: Applied Biosystems™ AM9914G
RNase-Free, exonuclease-free, endonuclease-free.
Nuclease-free water for use in RNA applications. 1.75 mL/tube
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Grade Molecular Biology
Packaging 1 tube(s)
Quantity 1 x 1.75 mL tube
Shipping Condition Room Temperature
Manuals
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2401037 Certificate TypeCertificate of Analysis Date27/01/2025 Product CodeAM9914G
Lot Number2302033 Certificate TypeCertificate of Analysis Date27/01/2025 Product CodeAM9914G
Lot Number2310036 Certificate TypeCertificate of Analysis Date25/01/2024 Product CodeAM9914G
Lot Number2307035 Certificate TypeCertificate of Analysis Date07/09/2023 Product CodeAM9914G
Lot Number2305034 Certificate TypeCertificate of Analysis Date21/07/2023 Product CodeAM9914G
For research use only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Applied Biosystems™ Nuclease-free Water (1.75 mL/tube) >