missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARA54 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | ARA54 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ARA54 Polyclonal antibody specifically detects ARA54 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
ARA54 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction, Zinc Finger | |
PBS, pH 7.2, 40% glycerol | |
9604 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Androgen receptor-associated protein 54, ARA54androgen receptor associated protein 54, EC 6.3.2.-, HFB30E3 ubiquitin-protein ligase RNF14, ring finger protein 14FLJ26004, TRIAD2, Triad2 protein | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TQLSALCKHLDNLWEEHRGSVVLFAWMQFLKEETLAYLNIVSPFELKIGSQKKVQRRTAQASPNTELDFGGAAGSDVDQEEIVDERAVQDVESLSNL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |