missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP30 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06084R
This item is not returnable.
View return policy
Description
ARHGAP30 Polyclonal specifically detects ARHGAP30 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ARHGAP30 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Rho GTPase activating protein 30 | |
Produced in rabbits immunized with E. coli-derived Human ARHGAP30 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
257106 | |
Store at 4°C in the dark. | |
IgG |