Learn More
Sartorius arium™ Water System Cartridge Set
Produce ultrapure water beyond the ASTM Type 1 standard. Sartorius arium™ Water System Cartridge Set is designed for installation in arium™ Pro, Basic or 611 systems. Each consumable kit includes a pre-treatment and polishing cartridge. Kits are specifically formulated for each particular system.
Brand:  Sartorius H2O-U-PACK
7013.30 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11747474
  • 7195.00 DKK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
In Stock
Add to basket
Alert:
*Tap water cartridges should be used only after the feed water specifications have been checked and following consultation with the responsible Sartorius™ application specialist.
Description
Description
arium™ water purification systems (Pro, Basic and/or 611 series) are designed to produce the highest purity laboratory water in the industry, with specifications that exceed the ASTM Type 1 standard. The cartridge sets are constructed of the highest quality materials with an advanced design to produce consistent, reproducible results.
  • Exceptional quality of the purification media
    • Optimized to remove organic and inorganic contaminants
    • Highly porous activated carbon combined with efficient ion exchange resins
    • Produces water with the lowest total organic carbon (TOC) content for systems using ultraviolet (UV) technology
  • Specially engineered so that water flows from top to bottom
    • The flow though the ion exchange medium prevents separation of the mixed-bed resin
    • Allows full use of each cartridge with no wasted media
    • Ensures a long cartridge life with unparalleled performance
  • Two-cartridge sets:
    • Pretreatment cartridge
    • Polishing cartridge
  • Specifically formulated for particular arium™ systems:
    • Analytical Kit for VF or UV systems
    • Biological Kit for UF systems
    • Environmental Kit for DI systems
    • Universal Kit for systems with tap water feed*
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM type 1
Pretreated
Ion Exchange, Activated Carbon
Standard, cell culture and chromatographic
Pretreatment, polishing cartridge
450 x 230 x 160 mm (WxDxH)
To 18.2 MΩ x cm ultrapure water in eq: 1.25
Polypropylene
Activated Carbon and Mixed Bed Ion Exchange Resin
arium™ Systems w/Tap Water Feed
Universal Kit
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Sartorius arium™ Water System Cartridge Set > Universal Kit