missing translation for 'onlineSavingsMsg'
Learn More
Arsenic, plasma standard solution, Specpure™ As 10,000μg/mL, Thermo Scientific Chemicals Product Code.: 11347927

Arsenic, plasma standard solution, Specpure™ As 10,000μg/mL, Thermo Scientific Chemicals

Product Code. 11347927
100 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11347927

Brand: Thermo Scientific Alfa Aesar 014369.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00085309

Specifications

Quantity 100 mL
UN Number UN3264
Physical Form Liquid
Concentration As in 5% HNO3
Grade Specpure™
Chemical Name or Material Arsenic plasma standard solution, As 10000μg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1550383 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number014369.AE,014369.AD,014369.AP
Lot Number1535594 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number014369.AE,014369.AD,014369.AP
Lot Number1504919 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number014369.AE,014369.AD,014369.AP
Lot Number1481462 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number014369.AE,014369.AD,014369.AP
Lot Number1474704 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog Number014369.AE,014369.AD,014369.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic, plasma standard solution, Specpure™ As 10,000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.