missing translation for 'onlineSavingsMsg'
Learn More

Artefenomel, MedChemExpress

MedChemExpress Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.

Encompass_Preferred

4683.12 DKK - 34856.53 DKK

Specifications

Chemical Name or Material Artefenomel
Physical Form Solid
CAS 1029939-86-3
Color Off-White
Purity Grade Notes Research
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16453207
Encompass_Preferred
View Documents
Medchem Express
HY-16762
50 mg
23405.54 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16437176
Encompass_Preferred
View Documents
Medchem Express
HY-16762
100 mg
34856.53 DKK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16429717
Encompass_Preferred
View Documents
Medchem Express
HY-16762
5 mg
4683.12 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16427188
Encompass_Preferred
View Documents
Medchem Express
HY-16762
10 mg
7805.20 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.
Specifications

Specifications

Artefenomel
1029939-86-3
Research
COVID-19-immunoregulation
DMSO : 4.2 mg/mL (8.94 mM; Need ultrasonic)
469.61
98.16%
Solid
Off-White
C28H39NO5
OZ439
[H][C@@]1(C[C@H](C2)C3)C4([C@@]3([H])C[C@H]2C1)O[C@]5(OO4)CC[C@H](CC5)C6=CC=C(C=C6)OCCN7CCOCC7
469.61
Research
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)

Recommended Storage : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Artefenomel, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.