Learn More
Artificial Seawater, Reagecon
High-quality, pure water product, presented in a convenient pack size.
Brand: Reagecon DSW25
This item is not returnable.
View return policy
Description
- Water Reference Standard, Water Reference Material
- Can be used as a matrix for a variety of solutes or as a cleaning/sanitization solution
Specifications
HDPE Container | |
25 L |