Learn More
RETSCH AS 200 Comfort Clamping Devices
8285.00 DKK - 9810.00 DKK
Specifications
Product Type | Clamping Devices |
---|---|
Diameter (Metric) | 100/150/200/203 mm |
For Use With (Equipment) | Analytical sieve shaker AS 200 |
Packaging Quantity | 1 Ea. |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
10379941
|
RETSCH
32.662.0004 |
Dry Sieving |
8285.00 DKK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
10330822
|
RETSCH
32.662.0006 |
Wet Sieving |
9810.00 DKK
Each |
Estimated Shipment: 24-10-2025
Log in to see stock. |
||||||
Specifications
Clamping Devices | |
100/150/200/203 mm | |
Analytical sieve shaker AS 200 | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
RETSCH AS 200 Comfort Clamping Devices