Learn More
Tocris Bioscience™ Asenapine maleate
Non-selective 5-HT, dopamine, adrenalin and histamine receptor antagonist; antipsychotic agent
Brand:  Tocris Bioscience™ 3737/50
4225.38 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18712514
  • 4765.00 DKK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Asenapine maleate is a novel psychopharmacologic agent. Displays antagonist activity at 5-HT, dopamine, noradrenalin and histamine receptor subtypes (pKi values are 8.60, 8.40, 10.15, 9.75, 10.46, 8.84, 9.60, 9.94, 8.85, 8.90, 8.84, 9.38, 8.95, 8.93, 8.9, 9.49, 8.91, 9.00 and 8.21 for 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT5A, 5-HT6, 5-HT7, D1, D2L, D2S, D3, D4, α1A, α2A, α2B, α2C, H1 and H2 receptors respectively). Displays no appreciable affinity for muscarinic receptors. Exhibits potent activity in animal models predictive of antipsychotic efficacy.
Specifications
Specifications
>98%
Asenapine maleate
Asenapine maleate
50 mg
401.84
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only