Learn More
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series
Aura™ Particulate Respirator 9300+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.
148.50 DKK - 19001.40 DKK
Specifications
Certifications/Compliance (Europe) | EN 149:2001 + A1:2009, CE Approved |
---|---|
Color | White |
Design Type | Flat-fold |
Disposable | Single-use |
Material | Polypropylene |
Product Code | Brand | Filter Type | Assigned Protection Factor (APF) | With Exhalation Valve | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Assigned Protection Factor (APF) | With Exhalation Valve | Quantity | Price | Quantity & Availability | |||||
16260262
|
3M™
7100138120 |
FFP2 | 10 | Yes | 5/Pk. | N/A | ||||||
17817603
|
3M™
7100138120 |
FFP2 | 10 | Yes | 1/Pk. |
1600.00 DKK
Pack of 40 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
16250262
|
3M™
7100138121 |
FFP2 | 10 | No | 5/Pk. | N/A | ||||||
16270262
|
3M™
7100138357 |
FFP3 | 20 | Yes | 5/Pk. | N/A | ||||||
16220262
|
3M™
7100134314 |
FFP3 | 20 | Yes | 10/Pk. |
800.00 DKK
Pack of 10 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
16200262
|
3M™
7100134313 |
FFP2 | 10 | Yes | 10/Pk. |
441.00 DKK
Pack of 10 |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16290252
|
3M™
7100134298 |
FFP2 | 10 | No | 20/Pk. | N/A | ||||||
16210262
|
3M™
7100134311 |
FFP3 | 20 | No | 20/Pk. | N/A | ||||||
16230262
|
3M™
7100139467 |
FFP2 | 10 | Yes | 330/Pk. | N/A | ||||||
16240262
|
3M™
7100138716 |
FFP3 | 20 | Yes | 330/Pk. | N/A | ||||||
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort compared to 3M™ Aura™ 9300+ equivalent model
- 3M™ Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are colour-coded (blue for FFP2, red for FFP3) for quick and easy identification
- Embossed upper panel reduces eyewear fogging
- Easy to store away with flat-fold design and individual wrapping
Specifications
EN 149:2001 + A1:2009, CE Approved | |
Flat-fold | |
Polypropylene | |
Standard | |
Non-sterile | |
No |
White | |
Single-use | |
General Purpose Particulate Respirator | |
Polyisoprene | |
Flat-fold |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series