Learn More
ratiolab™ Autoclavable Polypropylene Bags
This autoclavable, disposable waste bag is for the sterilization and disposal of hazardous and contaminated labware.
1510.00 DKK - 2430.00 DKK
Specifications
Product Type | Biohazard Disposal Bag |
---|---|
Autoclavable | Autoclavable |
Disposable | Single-use |
For Use With (Application) | Waste Disposal |
Includes | Bag with Indicator Patch |
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||||
13511030
|
ratiolab™
7002100 |
6 L | 300 x 500 mm |
1510.00 DKK
Pack of 500 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
15518713
|
ratiolab™
7002200 |
32 L | 800 x 600 mm |
2305.00 DKK
Pack of 500 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
15526939
|
ratiolab™
7002300 |
72 L | 1100 x 700 mm |
2430.00 DKK
Pack of 350 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
Description
- With BIOHAZARD warning Instruction and sterilization indicator patch
- Safety imprint in German, English, French and Spanish languages
- Rectangle white indicator field turns into dark when autoclaved
- For autoclaving, the bag must not be completely closed
- Autoclavable at 134°C for 20 min.
- Tolerance: ±5%
Specifications
Biohazard Disposal Bag | |
Single-use | |
Bag with Indicator Patch | |
50 μm |
Autoclavable | |
Waste Disposal | |
Polypropylene | |
2 mil |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
ratiolab™ Autoclavable Polypropylene Bags