Learn More
Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Package, 384-well, EnduraPlates
Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Package with EnduraPlates provides reliability and performance while enabling hand-free results.
Brand: Applied Biosystems™ A41200
64075.00 DKK valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Flexible modular design helps save space on a robotic deck
- The separate control panel module provides flexibility when integrating on a robotic deck
- The automated lid is compatible with programming for easy, hands-free operation with a liquid handler or plate stacker
- The heated cover slides forward to cover the plate nest, to help ensure thermal contact and eliminate evaporation
- The ATC is compatible with full-skirt or half-skirt plates
- Optimized for performance with Applied Biosystems instruments
- Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
- Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
- Available in multiple colors for easy organization of laboratory assays
- Excellent mechanical stability and flatness for your complete workflow needs
Specifications
±0.25°C (35°C to 99.9°C) | |
High-throughput Compatible | |
Standard | |
384-well, 20 μL full-skirted plate | |
No Display | |
Limited Warranty | |
USB, Wi-Fi, Ethernet | |
1 package | |
384-well Isothermal Block |
• Automated Thermal Cycler (ATC), 384-well, 1-meter cable • 5 x MicroAmp EnduraPlate Optical 384-well Clear Reaction Plates with Barcode (100 plates total) • 1 x MicroAmp Clear Adhesive Films (100 films) |
|
<0.5°C (20 sec. after reaching 95°C) | |
12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm) | |
100/240 V | |
Thermal Cycler | |
4°C to 105°C | |
2.8°C/sec. (Block), 1.6°C/sec. (Sample) | |
13.2 lb. (6.0 kg) |