Learn More
Hecht Karl™ Automatic Pipettes without Bottle
Brand:  Hecht Karl™ 41877018
Product Code. 10609431
  • 758.00 DKK / Each
Only null left
Estimated Shipment: 03-07-2025
Specifications
1 Each
5 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hecht Karl™ Automatic Pipettes without Bottle > Capacity: 5mL