missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 300 μL Universal Pipetter Tips

Fine-point tips fits popular brands of pipettes

226.27 DKK - 1587.38 DKK

Specifications

Volume (Metric) 1 to 300 μL
Color Clear
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Tip Style Micro Point, Universal
For Use With (Equipment) Universal Fit
View More Specs

Products 4
Product Code Brand Sterility Format Price Quantity & Availability  
Product Code Brand Sterility Format Price Quantity & Availability  
11354255
View Documents Promotion Details
Axygen™
T-350-C
Non-sterile Bulk
226.27 DKK
Pack of 1000
In Stock
Edge
Add to basket
 
12669535
View Documents
Axygen™
T-350-C-R-S
Sterile Rack
549.43 DKK
Pack of 960
Estimated Shipment: 30-09-2025
Add to basket
 
12083879
View Documents
Axygen™
T-350-C-R
Non-sterile Rack This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11384255
View Documents
Axygen™
T-350-C-R
Non-sterile Rack
465.15 DKK
Pack of 960
Estimated Shipment: 30-09-2025
Add to basket
 
Description

Description

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point

Order Info

  • Bulk: 1000 tips/pack, 240 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Specifications

1 to 300 μL
DNase, RNase, ATP, and Endotoxin Free
Universal Fit
Clear
Micro Point, Universal
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ 300 μL Universal Pipetter Tips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.