Learn More
Axygen™ AxyMats™ 96 Round Well Compression Mat for PCR Microplates
Suited to help reduce evaporation and ensure proper seal when used in conjunction with Axygen PCR Microplates and PlateMax™ sealing films
Brand:  Axygen™ CM-96-RD
Product Code. 11386274
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Axygen™ Compression AxyMatsTM are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microplates and PlateMaxTM sealing films. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
  • For use with 96/384 well and PCR microplates
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Natural
    Sterile
    96
    10/Pk.
    Round
    Silicone
    Autoclavable
    50/Cs.
    Compression Mat
    96/384-well PCR Microplate