Learn More
Axygen™ AxyMats™ Sealing Mat for 96 Well PCR Microplates
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand:  Axygen™ AM-96-PCR-RD
Product Code. 12142570
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Non-sterile
96
10/Pk.
Round
Silicone
Autoclavable
50/Cs.
Sealing Mat
96-well PCR Microplate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Axygen™ AxyMats™ Sealing Mat for 96 Well PCR Microplates > 0.2 μL; Maxyclear