Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
Designed to accommodate all multichannel pipettor
631.56 DKK - 2055.00 DKK
Specifications
Certifications/Compliance | ANSI, RNase/DNase-free, non-pyrogenic |
---|---|
Color | White |
Material | Polystyrene |
Type | Reagent Reservoir |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Packaging | Price | Quantity & Availability | ||||
12537967
![]() |
Axygen™
RES-2CV-25 |
25 mL | Non-sterile | Divided; 100/Cs. |
688.89 DKK
Pack of 100 |
In Stock
Log in to see stock. |
||||
12537997
![]() |
Axygen™
RES-2CV-25-S |
25 mL | Sterile | Divided; 200/Cs. |
1314.88 DKK
Pack of 200 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
12547967
![]() |
Axygen™
RES-2CV-25-SI |
25 mL | Sterile | Divided; Individually Wrapped; 100/Cs. |
1680.00 DKK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
12547997
![]() |
Axygen™
RES-V-100-S |
100 mL | Sterile | 200/Cs. |
1567.25 DKK
Pack of 200 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
12567967
![]() |
Axygen™
RES-V-100-SI |
100 mL | Sterile | Individually Wrapped; 100/Cs. |
2042.31 DKK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
12557967
![]() |
Axygen™
RES-V-100 |
100 mL | Non-sterile | 100/Cs. |
631.56 DKK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
10241570
![]() |
Axygen™
RES-V-25 |
25 mL | Non-sterile | 100/Cs. |
1050.00 DKK
Pack of 100 |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
12557997
![]() |
Axygen™
RES-V-25-S |
25 mL | Sterile | 200/Cs. |
1905.00 DKK
Pack of 200 |
In Stock
Log in to see stock. |
||||
12587967
![]() |
Axygen™
RES-V-25-SI |
25 mL | Sterile | Individually Wrapped; 100/Cs. |
1775.00 DKK
Pack of 100 |
Estimated Shipment: 24-09-2025
Log in to see stock. |
||||
12597967
![]() |
Axygen™
RES-V-50 |
50 mL | Non-sterile | 100/Cs. |
821.00 DKK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
12567997
![]() |
Axygen™
RES-V-50-S |
50 mL | Sterile | 200/Cs. |
2055.00 DKK
Pack of 200 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
12507977
![]() |
Axygen™
RES-V-50-SI |
50 mL | Sterile | Individually Wrapped; 100/Cs. |
1849.14 DKK
Pack of 100 |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||||
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
ANSI, RNase/DNase-free, non-pyrogenic | |
Polystyrene | |
Single-use |
White | |
Reagent Reservoir |