Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
GREENER_CHOICE
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand:  Axygen™ HS-150-QPCR
 View more versions of this product
Product Code. 12322099
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Peelable Heat Sealing Film for qPCR
PlateMax Plate Sealer
500/Cs.
Plate Sealing
Polyester
100/Pk.