missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

For sample collection, in-vitro growth chambers and long term storage.

Brand:  Axygen™ P-384-240SQ-C-S

 View more versions of this product

Product Code. 11310115

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    ANSI, SLAS 1-2004
    Without Lid
    Clear
    240 μL
    Sterile
    Sample collection, in-vitro growth chambers and long term storage
    50/Cs.
    -112°F
    Standard
    16 x 24
    Polypropylene
    384
    Square
    4000 x g
    5/Pk.
    -80°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Axygen™ Storage Microplates > 384-well; 240 μL; Yes; Square

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.