missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelaic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼102°C |
Boiling Point | 286°C (100mmHg) |
Beilstein | 1101094 |
Merck Index | 14,905 |
Solubility Information | Soluble in water (2.4mg/ml at 20°C),ethanol (5%),DMSO,methanol (70mg/ml),and alcohol. |
Formula Weight | 188.22 |
Percent Purity | 96% |
Density | 1.225 g/mL |
Quantity | 25 g |
Flash Point | 215°C (419°F) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'