missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Screwcaps for Reagent Bottles

Phenolic cap with PTFE faced sealing disc for chemical resistance

Brand:  Pyrex 4503/60

Product Code. 12633292

  • 1065.00 DKK / Pack of 5
Only null left
Estimated Shipment: 17-10-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Phenolic cap with PTFE faced sealing disc for chemical resistance
  • Autoclavable up to 121°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
PTFE faced sealing disc of Ethylene Propylene
5/PK
PTFE faced
Reagent and media bottles, culture tubes
Screw cap
Phenolic Resin
60mm
Autoclavable
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ Screwcaps for Reagent Bottles > SVL 60mm thread size

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.