Learn More
Azlon™ Silicone Rubber Hand/Finger Protector
598.50 DKK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Use promo code "23259" to get your promotional price.
Click to view available options
Size:
Finger
Medium
Small
Description
- Capable of withstanding temperatures up to 250°C
- For handling cold objects down to -57°C
- Contact surfaces are multi-studded for positive grip
Specifications
Material | Silicone |
Product Type | Hand/Finger Protector |
Size | Medium |
For Use With (Application) | For Safe Handling Of Hot Beakers, Flasks etc |