Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Printable, polyester labels for Thermal Transfer Printers.
Brand:  Brady™ 030514
 View more versions of this product
Product Code. 11528264
  • 1020.00 DKK / 1 roll
In Stock
Explore more special offers
Description
  • Permanent adhesive
  • For smooth surfaces
Specifications
Specifications
Autoclavable
Permanent Acrylic
Gloss
Thermal Transfer
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling
1
Rectangle
0.076 mm
22.9 mm
6.4 mm
THT-49-423-10
UL, CSA and AGA Approved
White
Polyester (B-423)
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
10,000 Labels
-70°C to +110°C
8.9 mm
22.86 mm
6.4 mm
Product Title
Brady™ B-423 Polyester Thermal Transfer Labels > Polyester (B-423) Thermal Transfer Labels; Color: White; Height (Metric): 6.4 mm; Labels per Row: 1; Vertical Repeat (Metric): 8.9 mm; Width (Metric): 22.9 mm