Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Printable, polyester labels for Thermal Transfer Printers.
Brand:  Brady™ 030509
 View more versions of this product
Product Code. 11578254
  • 1265.00 DKK / 1 roll
In Stock
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Permanent adhesive
  • For smooth surfaces
Specifications
Specifications
Autoclavable
Permanent Acrylic
Gloss
Thermal Transfer
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling
1
Rectangle
0.076 mm
34.9 mm
6.4 mm
THT-44-423-10
UL, CSA and AGA Approved
White
Polyester (B-423)
Non-sterile
BBP™81, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
10,000 Labels
-70°C to +110°C
8.9 mm
34.93 mm
6.4 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ B-423 Polyester Thermal Transfer Labels > Polyester (B-423) Thermal Transfer Labels; Color: White; Height (Metric): 6.4 mm; Labels per Row: 1; Vertical Repeat (Metric): 8.9 mm; Width (Metric): 34.9 mm