Få mere at vide
Tocris Bioscience™ B I09
Beskrivelse
B I09 is an IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.
Tekniske data
Tekniske data
| CAS | 1607803-67-7 |
| Mængde | 50 mg |
| Opløselighed | Soluble to 50mM in DMSO |
| Molekylær formel | C16H17NO5 |
| Formel vægt | 303.31g/mol |
| Farve | Beige |
| Produkttype | Inhibitor |
For Research Use Only