Learn More
Brady™ B30 Series Continuous Vinyl Labels
Labels constructed of B-7569 Vinyl for indoor/outdoor use.
Brand:  Brady™ 120913
 View more versions of this product
Product Code. 15843975
  • 908.00 DKK / Each
Explore more special offers
Description
Description
  • B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels, and visual workplace labels
Specifications
Specifications
Permanent Acrylic
Gloss
Thermal Transfer
30 m x 29 mm
Orange
Vinyl (B-7569)
BBP30, BBP31, BBP33, BBP35, BBP37, i3300, i5300, S3000, S3100