Learn More
Motic™ BA210E Trinocular Compound Microscope
Use this microscope for both educational and teaching environments. Motic™ BA210E Trinocular Compound Microscope is ideal for a variety of life sciences and medical applications.
Brand: Motic™ 1100100403761
7571.49 DKK valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Excellent spherical aberration correction to significantly improve field flatness and resolution.
- Superior color fidelity through new multi-layer coated lenses.
- Significantly increased working distances and lead-free manufacturing process according to RoHS standard.
- The ELITE model's addition of a new rackless stage concept without prominent gear rack allows a more convenient use of the x/y movement.
- Full freedom of Illumination options, the new model allows for easy interchangeability of its 6V/30W halogen bulb with LED modules
- Motic Infinity Optics (CCIS™) allows a higher level of performance in education and training.
- Contrast methods like phase contrast, polarization and dark field are easily performed using optional accessories.
Specifications
100/240 V | |
CCIS EC Plan | |
Universal power supply 100/240 V (CE) | |
Trinocular compound microscope | |
High Eyepoint N-WF 10x/20 mm with diopter adjustment on both eyepieces |
4X, 10X, 40XS, 100XS (oil) | |
Reversed quadruple revolving nosepiece. | |
Focusable Abbe condenser N.A. 1.25 with iris diaphragm and slot | |
Halogen with Intensity Control 6V/30w |