Learn More
Gibco™ Bacto™ Malt Extract
The water-soluble portion of malted barley.
670.00 DKK - 7650.00 DKK
Specifications
Form Powder
Source Animal origin-free, malted barley
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16289841
View Documents Promotion Details
Gibco™
218630
500 g
670.00 DKK
500g
View alternative products
Save Up to 
»
16279841
View Documents Promotion Details
Gibco™
218610
10 kg
7650.00 DKK
10kg
View alternative products
Save Up to 
»
Description
Description
  • Bacto Malt Extract is the water-soluble portion of malted barley
  • Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium
  • It has been used successfully in microbial fermentation applications
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Specifications
Powder
Animal origin-free, malted barley
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 1460 Days