Learn More
Tocris Bioscience™ BAG 956
Dual PI 3-kinase and PDPK1 (PDK1) inhibitor
Brand:  Tocris Bioscience™ 3606/50
 View more versions of this product
Product Code. 18024652
  • 6750.00 DKK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
BAG 956 is a dual PDPK1 (PDK1) and class I PI 3-K inhibitor (IC50 values are 245, 56, 446, 35 and 117 nM for PDPK1 and PI 3-K p110 -α, -β, -δ, and -γ respectively). Inhibits cellular AKT phosphorylation at Thr308. Blocks cell proliferation, causing arrest in G1 phase of the cell cycle. Slows tumor progression in mouse xenograft models.
Specifications
Specifications
98%
50 mg
427.5
Store at -20°C
BAG 956
BAG 956
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only