Learn More
Bar Magnets
POA
Specifications
Product Type Magnets
Height (Metric) 6 mm
Quantity 1 Pk.
View More Specs
Products 2
Product Code Brand Width (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Price Quantity & Availability  
12443120
Encompass
View Documents
N/A
50225
75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12463120
Encompass
View Documents
N/A
50227
150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
Magnets
6 mm
1 Pk.
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.