Learn More
Thermo Scientific™ Barnstead™ MegaPure™ ACS
Control the operation of your Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Still with this automatic collection system, which collects up to 45L of water.
Brand:  Thermo Scientific™ B440704
 View more versions of this product
Product Code. 12714846
  • 28392.37 DKK / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Control the operation of your glass still with the Thermo Scientific™ Barnstead™ MegaPure™ ACS (Automatic Collection System), which collects up to 45L of water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Automatic Collection System (ACS) Unit
Automatic Collection System (ACS)
16 in.
16 in.
38 in.
Mega-Pure Glass Still
16 x 16 x 38 in. (41 x 41 x 97 cm)
41 cm
41 cm
97 cm